Thermo Scientific Chemicals

Di-n-butyltin dilaurate, 95%, Thermo Scientific Chemicals

Catalog number: 071130.36
500 g, Each
Thermo Scientific Chemicals

Di-n-butyltin dilaurate, 95%, Thermo Scientific Chemicals

Catalog number: 071130.36
500 g, Each
Quantity
Catalog number: 071130.36
also known as 071130-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
77-58-7
IUPAC Name
dibutyltin dilaurate
Molecular Formula
C32H66O4Sn
InChI Key
XWVGXTYHPCKSLV-UHFFFAOYSA-N
SMILES
CCCC[SnH2]CCCC.CCCCCCCCCCCC(O)=O.CCCCCCCCCCCC(O)=O
Appearance (Color)
Clear pale yellow to yellow
Assay (unspecified)
17.7 to 18.9 % Total Tin
Chloride content
0.15 wt%, max
Water Content (Karl Fischer Titration)
0.5 %, max
Form
Liquid

Description

Di-n-butyltin dilaurate is used as a catalyst for the production of polyurethanes as well as for the transesterification reactions. It is involved in the vulcanization of silicones and a stabilizer in polyvinyl chloride (PVC). It acts as a rust inhibitor for polyurethanes, polyols, silicones and as a fuel additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text