Thermo Scientific Chemicals
Tetra-n-propyltin, Thermo Scientific Chemicals
Catalog number: 071137.09
10 g, Each
Thermo Scientific Chemicals
Tetra-n-propyltin, Thermo Scientific Chemicals
Catalog number: 071137.09
10 g, Each
Quantity
Catalog number: 071137.09
also known as 071137-09
Price (USD)
Quantity
-
Have Questions?
Description
It can be used to produce Dichlor-dipropyl-stannan and propane-1-sulfinyl chloride at room temperature. Applications include preservation of wood, textiles, leather, and paper.