Thermo Scientific Chemicals

Tetraethyltin, 98%, Thermo Scientific Chemicals

Catalog number: 071144.18
50 g, Each
Thermo Scientific Chemicals

Tetraethyltin, 98%, Thermo Scientific Chemicals

Catalog number: 071144.18
50 g, Each
Quantity
Catalog number: 071144.18
also known as 071144-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
597-64-8
IUPAC Name
tetraethylstannane
Molecular Formula
C8H20Sn
InChI Key
RWWNQEOPUOCKGR-UHFFFAOYSA-N
SMILES
CC[Sn](CC)(CC)CC
Assay (GC)
> 97.0%

Description

Tetraethyltin is used in the electronics industry. Used as catalyst for olefin polymerization and to produce other organotin compounds. Also used as a preservative for wood, textiles, paper, and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text