Thermo Scientific Chemicals

Trimethyltin chloride, Thermo Scientific Chemicals

Catalog number: 071166.09
10 g, Each
Thermo Scientific Chemicals

Trimethyltin chloride, Thermo Scientific Chemicals

Catalog number: 071166.09
10 g, Each
Quantity
Catalog number: 071166.09
also known as 071166-09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1066-45-1
Form
Crystalline solid
Appearance (Color)
White
Assay (GC)
>98+%

Description

Trimethyltin chloride is used as a heat stabilizer in the manufacture of vinyl window blinds. It is also used as a stabilizer for plastics and as additive to paint, as antifouling agent. It also serves as a byproduct of methyltin manufacture. Further, it is used as a precursor to vinyltrimethylstannane and in polyvinyl chloride stabilization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text