Thermo Scientific Chemicals

Trimethyltin hydroxide, 98%, Thermo Scientific Chemicals

Catalog number: 071167.06
5 g, Each
Thermo Scientific Chemicals

Trimethyltin hydroxide, 98%, Thermo Scientific Chemicals

Catalog number: 071167.06
5 g, Each
Quantity
Catalog number: 071167.06
also known as 071167-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
56-24-6
IUPAC Name
trimethylstannanol
Molecular Formula
C3H10OSn
InChI Key
OJZNYUDKNVNEMV-UHFFFAOYSA-M
SMILES
C[Sn](C)(C)O
Form
Solid
Appearance (Color)
White to off-white
Assay from Suppliers CofA
≥97.5%

Description

Trimethyltin hydroxide is used in the synthesis of modified thymines, which is useful as inhibitors of RNase A. It is also used in the preparation of ketoheterocycle inhibitors of fatty acid amide inhibitors. It reacts with 2,2-disubstituted benzothiazolines to form trialkyltin derivatives of sulphur containing Schiff base complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text