Thermo Scientific™

Dionex™ AutoTrace™ 280 Solid-Phase Extraction Instrument

Catalog number: 071385
Have Questions?
Thermo Scientific™

Dionex™ AutoTrace™ 280 Solid-Phase Extraction Instrument

Catalog number: 071385
Have Questions?

Extract analytes of interest from large volume aqueous samples easily and efficiently with the Thermo Scientific™ Dionex™ AutoTrace™ 280 Solid-Phase Extraction (SPE) Instrument. The AutoTrace 280 automates all four steps of SPE (conditioning, loading, rinsing, and eluting), reducing solvent consumption and improving recovery and reproducibility. Boost productivity and reduce cost of analysis by simultaneously processing up to six samples in 2-3 hours with only 15 minutes of operator involvement. The AutoTrace 280 meets requirements in US EPA methods including the 500 and 600 series.

 
Catalog Number
071385
Unit Size
Each
For Use With (Equipment)
Cartridges with 6 mL Plungers
Price (USD)
Full specifications
DescriptionFor cartridges with 6 mL plungers
Frequency47 to 63 Hz
PressureOutput: 0 to 30 psi (0 to 1.4 bars), Input: 100 psi (6.9 bars) maximum
Dimensions (D x W x H)Instrument: 57 x 63.5 x 69 cm
For Use With (Equipment)Cartridges with 6 mL Plungers
TypeAutoTrace 280 SPE System
Voltage100/240 V ±10%
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
47 mm Disks
Price (USD)
Full specifications
DescriptionFor 47 mm disks
Frequency47 to 63 Hz
PressureOutput: 0 to 30 psi (0 to 1.4 bars), Input: 100 psi (6.9 bars) maximum
Dimensions (D x W x H)Instrument: 57 x 63.5 x 69 cm
For Use With (Equipment)47 mm Disks
TypeAutoTrace 280 SPE System
Voltage100/240 V ±10%
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
Cartridges with 1 mL Plungers
Price (USD)
Full specifications
DescriptionFor cartridges with 1 mL plungers
Frequency47 to 63 Hz
PressureOutput: 0 to 30 psi (0 to 1.4 bars), Input: 100 psi (6.9 bars) maximum
Dimensions (D x W x H)Instrument: 57 x 63.5 x 69 cm
For Use With (Equipment)Cartridges with 1 mL Plungers
TypeAutoTrace 280 SPE System
Voltage100/240 V ±10%
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
Cartridge with 3 mL Plungers
Price (USD)
Full specifications
DescriptionFor cartridges with 3 mL plungers
Frequency47 to 63 Hz
PressureOutput: 0 to 30 psi (0 to 1.4 bars), Input: 100 psi (6.9 bars) maximum
Dimensions (D x W x H)Instrument: 57 x 63.5 x 69 cm
For Use With (Equipment)Cartridge with 3 mL Plungers
TypeAutoTrace 280 SPE System
Voltage100/240 V ±10%
Unit SizeEach
Catalog Number
Unit Size
Each
For Use With (Equipment)
Cartridge with 6 mL Glass Plungers
Price (USD)
Full specifications
DescriptionFor cartridges with 6 mL glass plungers
Frequency47 to 63 Hz
PressureOutput: 0 to 30 psi (0 to 1.4 bars), Input: 100 psi (6.9 bars) maximum
Dimensions (D x W x H)Instrument: 57 x 63.5 x 69 cm
For Use With (Equipment)Cartridge with 6 mL Glass Plungers
TypeAutoTrace 280 SPE System
Voltage100/240 V ±10%
Unit SizeEach
Showing 5 of 5
Catalog NumberSpecificationsUnit SizeFor Use With (Equipment)Price (USD)
071385Full specifications
EachCartridges with 6 mL PlungersRequest A Quote
DescriptionFor cartridges with 6 mL plungers
Frequency47 to 63 Hz
PressureOutput: 0 to 30 psi (0 to 1.4 bars), Input: 100 psi (6.9 bars) maximum
Dimensions (D x W x H)Instrument: 57 x 63.5 x 69 cm
For Use With (Equipment)Cartridges with 6 mL Plungers
TypeAutoTrace 280 SPE System
Voltage100/240 V ±10%
Unit SizeEach
071386Full specifications
Each47 mm DisksRequest A Quote
DescriptionFor 47 mm disks
Frequency47 to 63 Hz
PressureOutput: 0 to 30 psi (0 to 1.4 bars), Input: 100 psi (6.9 bars) maximum
Dimensions (D x W x H)Instrument: 57 x 63.5 x 69 cm
For Use With (Equipment)47 mm Disks
TypeAutoTrace 280 SPE System
Voltage100/240 V ±10%
Unit SizeEach
072604Full specifications
EachCartridges with 1 mL PlungersRequest A Quote
DescriptionFor cartridges with 1 mL plungers
Frequency47 to 63 Hz
PressureOutput: 0 to 30 psi (0 to 1.4 bars), Input: 100 psi (6.9 bars) maximum
Dimensions (D x W x H)Instrument: 57 x 63.5 x 69 cm
For Use With (Equipment)Cartridges with 1 mL Plungers
TypeAutoTrace 280 SPE System
Voltage100/240 V ±10%
Unit SizeEach
072605Full specifications
EachCartridge with 3 mL PlungersRequest A Quote
DescriptionFor cartridges with 3 mL plungers
Frequency47 to 63 Hz
PressureOutput: 0 to 30 psi (0 to 1.4 bars), Input: 100 psi (6.9 bars) maximum
Dimensions (D x W x H)Instrument: 57 x 63.5 x 69 cm
For Use With (Equipment)Cartridge with 3 mL Plungers
TypeAutoTrace 280 SPE System
Voltage100/240 V ±10%
Unit SizeEach
072606Full specifications
EachCartridge with 6 mL Glass PlungersRequest A Quote
DescriptionFor cartridges with 6 mL glass plungers
Frequency47 to 63 Hz
PressureOutput: 0 to 30 psi (0 to 1.4 bars), Input: 100 psi (6.9 bars) maximum
Dimensions (D x W x H)Instrument: 57 x 63.5 x 69 cm
For Use With (Equipment)Cartridge with 6 mL Glass Plungers
TypeAutoTrace 280 SPE System
Voltage100/240 V ±10%
Unit SizeEach
Showing 5 of 5
Improved reliability and recovery
  • Automated SPE minimizes operator intervention
  • Positive pressure loading and elution of samples and solvents provide constant flow for improved analytical precision

Flexibility in operation
  • Loads sample volumes from 20 mL to 4 L
  • Compatible with 1, 3, and 6 mL cartridges and with 47 mm SPE disks
  • Automatically conditions, rinses, and elutes SPE cartridges with a choice of five solvents
  • Offers a choice of six different collection vial racks

Improved laboratory safety
  • Minimal sample handling reduces operator exposure to hazardous chemicals
  • No hood required

Enhanced laboratory productivity and reduced costs
  • Unattended operation
  • Batch processing of up to 6 samples
  • Reduced solvent usage
  • Segregates aqueous and solvent waste using separate waste lines for efficient, cost-effective waste handling

Supports a range of matrices and applications
  • Ability to process samples containing suspended solids
  • Meets requirements for solid-phase extraction for U.S. EPA methods including 500 and 600 series

Recommended use
  • Analyze prepared samples with Thermo Scientific LC or GC systems for a fully integrated workflow
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text