Search Thermo Fisher Scientific
Remove ionic contaminants from eluents continuously without offline chemical regeneration using Dionex CR-TC Continuously Regenerated Trap columns.
Catalog Number | Description | For Use With (Equipment) |
---|---|---|
072079 | Dionex™ CR-CTC (Capillary) Continuously Regenerated Cation Trap Column | Standard Cation Capillary RFIC Systems |
060477 | Dionex™ CR-ATC Continuously Regenerated Anion Trap Column | Standard Anion Capillary RFIC Systems up to 3,000 psi |
072078 | Dionex™ CR-ATC (Capillary) Continuously Regenerated Anion Trap Column | Standard Anion Capillary RFIC Systems |
075550 | Dionex™ CR-ATC 500 Continuously Regenerated Anion Trap Column | All Anion, RFIC, and HPIC Systems that are Not Consumables Monitoring Enabled |
066262 | Dionex™ CR-CTC II Continuously Regenerated Cation Trap Column | Standard RFIC Systems That Operate up to 3,000 psi |
088662 | Dionex™ CR-ATC 600 Continuously Regenerated Anion Trap Column | Dionex ICS-6000 and Dionex Integrion Systems |
088663 | Dionex™ CR-CTC 600 Continuously Regenerated Cation Trap Column | Dionex ICS-6000 and Dionex Integrion Systems |
075551 | Dionex™ CR-CTC 500 Continuously Regenerated Cation Trap Column | Dionex HPIC Systems that are Not Consumables Monitoring Enabled |
104-60001 | Dionex™ CR-CTC III Continuously Regenerated Cation Trap Column | Dionex ICS-6000 and Dionex Integrion Systems |
Remove ionic contaminants from eluents continuously, without the need for offline chemical regeneration. Thermo Scientific™ Dionex™ CR-TC Continuously Regenerated Trap Columns are electrolytically regenerated trap columns specifically designed for Reagent-Free™ ion chromatography (RFIC™) systems. When plumbed after the Thermo Scientific™ Dionex™ EGC Eluent Generation Cartridge, the Dionex CR-TC trap column removes all ionic contaminants from the deionized feed water, providing low drift during gradient operations.
Dionex CR-TC columns:
Which Dionex CR-TC column to use with which IC system:
Dionex CR-CTC III Continuously Regenerated Cation Trap columns are high-pressure electrolytically regenerated trap columns designed for use with eluent generator devices and compatible with Dionex Integrion and Dionex ICS-6000 HPIC systems
The Dionex CR-CTC III offers: