Thermo Scientific™

Q Exactive™ BioPharma Platform

Catalog number: 0726055
Have Questions?
Thermo Scientific™

Q Exactive™ BioPharma Platform

Catalog number: 0726055
Have Questions?

Realize complete BioPharma characterization with the Thermo Scientific™ Q Exactive™ BioPharma platform, which offers distinct operational modes optimized for the top three protein characterization workflows:

  • High Mass Range mode offers optimized intact protein analysis under both native and denaturing conditions providing the highest quality spectra.
  • Protein mode ensures isotopic resolution of subunits and and facilitates top/middle-down sequencing.
  • Standard mode performs peptide mapping with unparalleled acquisition speed, mass accuracy, and spectral quality.
 
Catalog Number
0726055
Unit Size
Each
Scan Rates
Up to 12 Hz
Resolution
280,000 at m/z 200
Mass Range
50 to 8000 m/z
Price (USD)
Full specifications
TypeBioPharma Platform
Dynamic Range>5000:1
Mass Range50 to 8000 m/z
Mass AccuracyInternal: <1 ppm RMS, External: <3 ppm RMS under defined conditions
Resolution280,000 at m/z 200
Scan RatesUp to 12 Hz
Polarity SwitchingOne Full Cycle in <1 Sec (One Full Positive Mode Scan and One Full Negative Mode Scan (at a Resolution Setting of 35,000))
SensitivityFull MS: 500 fg Buspirone on Column S/N 100:1
SIM: 30 fg Buspirone on Column S/N 100:1
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeScan RatesResolutionMass RangePrice (USD)
0726055Full specifications
EachUp to 12 Hz280,000 at m/z 20050 to 8000 m/zRequest A Quote
TypeBioPharma Platform
Dynamic Range>5000:1
Mass Range50 to 8000 m/z
Mass AccuracyInternal: <1 ppm RMS, External: <3 ppm RMS under defined conditions
Resolution280,000 at m/z 200
Scan RatesUp to 12 Hz
Polarity SwitchingOne Full Cycle in <1 Sec (One Full Positive Mode Scan and One Full Negative Mode Scan (at a Resolution Setting of 35,000))
SensitivityFull MS: 500 fg Buspirone on Column S/N 100:1
SIM: 30 fg Buspirone on Column S/N 100:1
Unit SizeEach
Showing 1 of 1

The Thermo Scientific™ Q Exactive™ Plus, Q Exactive HF and Q Exactive HF-X Hybrid Quadrupole-Orbitrap™ mass spectrometers add superior denatured and native MS intact analysis and subunit top/middle-down analysis capabilities to the most powerful benchtop peptide mapping instruments available. When combined with Thermo Scientific™ BioPharma Finder™ software it provides a complete integrated hardware and software solution for BioPharma characterization.

The Thermo Scientific™ Q Exactive™ BioPharma platform offers distinct operational modes that have been optimized for the top 3 protein characterization workflows:

  • High Mass Range (HMR) mode was designed specifically for intact therapeutic protein workflows with the aim of providing ease of use an flexibility as needed. HMR mode is optimized for both conventional intact monoclonal antibody analysis performed in denaturing conditions, as well as native MS analysis which is ideal for antibody drug conjugates (ADCs) and other heterogeneous antibody samples.
  • Protein mode on the Q Exactive HF is a powerful combination that has set the standard for the highest quality isotopically resolved data achievable on a benchtop mass spectrometer. Deconvolution of subunit MS spectra is easy and powerful with Thermo Scientific™ BioPharma Finder™ Software.
  • Peptide mapping with Standard mode is state-of-the-art with the Q Exactive BioPharma, which is a culmination of decades of leadership in the development of data dependent acquisition (DDA).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text