Search Thermo Fisher Scientific
Catalog Number | Description |
---|---|
072629 | Electrolytic Water Purifier Kit for Anion Analysis |
071553 | Electrolytic Water Purifier Replacement Unit (EWP only) for Anion Analysis |
071859 | Electrolytic Water Purifier Replacement Unit (EWP only) for Cation Analysis |
072630 | Electrolytic Water Purifier Kit for Cation Analysis |
Effluent from the conductivity cell supplies feed water for the Electrolytic Water Purifier. Water passing through the Electrolytic Water Purifier's main purification chamber exits the device free of ionic contamination, including carbonate. This device is used in AutoPrep mode, which enhances simplicity by eliminating the need for an auxiliary loading pump and removing potential contamination from the pump.
The Electrolytic Water Purifier is powered by either the:
The Electrolytic Water Purifier is one of several examples of Reagent-Free™ Ion Chromatography - Electrolytic Sample Preparation (RFIC™-ESP™).