1x1 image pixel for data collection
Dionex AS-DV Autosampler Preventive Maintenance Kit
Product Image
Thermo Scientific™
Dionex AS-DV Autosampler Preventive Maintenance Kit
Perform routine maintenance on Dionex™ AS-DV™ autosampler and ensure instrument uptime and maximum productivity with this convenient preventive maintenance kit.
Have Questions?
Catalog number 072678
Price (USD)
562.00
Each
Add to cart
Price (USD)
562.00
Each
Add to cart
Safeguard your lab productivity and protect your investment by performing routine maintenance on your Thermo Scientific™ Dionex™ AS-DV™ autosampler with the help of this preventive maintenance kit. Preventive care is critical to the life and availability of your autosampler and is recommended on an annual basis. Without regular maintenance, unattended, lingering problems can decrease instrument performance and lead to unnecessary and unplanned service costs.
The Dionex AS-DV preventive maintenance kit includes the following items:
  • Preventive maintenance manual
  • Dionex AS-DV sampling tip assembly
If needed, rebuild kits for optional AS-DV high pressure valves and lubricant must be ordered separately.
Specifications
TypeMaintenance Kit
IncludesPreventive maintenance manual, Dionex AS-DV sampling tip assembly
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link