Terbium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Terbium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Terbium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Terbium(III) nitrate hydrate, H2N3O10Tb, CAS Number-94219-56-4, terbium 3+ hydrate trinitrate, 3no3.tb.h2o, 25g, 99.9% (REO), Hazard Class: 5.1; Packaging Group: III, 233-138-7, 344.94 (anhydrous), H272-H315-H319-H335
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 074103.14
also known as 074103-14
Price (USD)/ Each
243.65
Online exclusive
270.00 
Save 26.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
243.65
Online exclusive
270.00 
Save 26.35 (10%)
Add to cart
Terbium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Catalog number074103.14
Price (USD)/ Each
243.65
Online exclusive
270.00 
Save 26.35 (10%)
-
Add to cart
Chemical Identifiers
CAS94219-56-4
IUPAC Nameterbium(3+) trinitrate
Molecular FormulaN3O9Tb
InChI KeyYJVUGDIORBKPLC-UHFFFAOYSA-N
SMILES[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormWet Crystalline Aggregates
Total Rare Earth Oxide Impurities≤0.1% (1000 ppm)
Appearance (Color)Pale green
Assay (special)40.5-42.1% Tb₄O₇
Terbium(III) nitrate hexahydrate forms a microporous material, which is used for sensor applications due to its luminescent properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only