Quickly remove or extract polar impurities from water and other liquids using the Thermo Scientific™ Dionex™ SolEx™ Unbonded Silica (Acid Washed) Silica-Based SPE Cartridges. This unbonded silica SPE cartridge is designed for use with the Dionex™ AutoTrace™ 280 SPE system or other SPE instrumentation such as vacuum manifolds. They are available in 6 mL cartridges with 0.5 g of packing.
About Dionex SolEx SPE Cartridges SolEx SPE Cartridges extract organic pollutants from a liquid sample off-line using a solid-phase adsorbent. The cartridges employ a variety of chemistries depending on the application. The silica- or carbon-based absorbents are bound or encapsulated in a membrane filter (called disk format) or packed in a cartridge (syringe bodies). Water (ground, waste, and drinking) is the most common liquid sample extracted, but these SPE cartridges are also used to extract contaminants from milk, fruit juices, urine, and other matrices. These cartridges are designed for use with Dionex AutoTrace 280 SPE system or other SPE instrumentation such as vacuum manifolds.
- Sorbant material made of irregular bonded silica particles
- Excellent capacity from packed with 40 to 63 micron particles
- Long 24-month shelf life
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.