Thermo Scientific Chemicals

Titanium(IV) isopropoxide, 97+%, Thermo Scientific Chemicals

Catalog number: 077115.57
4 x 500 g, Each
Thermo Scientific Chemicals

Titanium(IV) isopropoxide, 97+%, Thermo Scientific Chemicals

Catalog number: 077115.57
4 x 500 g, Each
Quantity
Catalog number: 077115.57
also known as 077115-57
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
546-68-9
IUPAC Name
titanium(4+) tetrakis(propan-2-olate)
Molecular Formula
C12H28O4Ti
InChI Key
VXUYXOFXAQZZMF-UHFFFAOYSA-N
SMILES
[Ti+4].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Form
Liquid
Color (APHA)
≤25
Assay (unspecified)
Ti: 27.50-28.30% (as TiO2)
Chloride content
≤100ppm
Specific Gravity
0.950-0.960 @ 25?C

Description

Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids.Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text