Thermo Scientific Chemicals

Titanium(IV) n-butoxide, 99+%, Thermo Scientific Chemicals

Catalog number: 077124.09
10 g, Each
Thermo Scientific Chemicals

Titanium(IV) n-butoxide, 99+%, Thermo Scientific Chemicals

Catalog number: 077124.09
10 g, Each
Quantity
Catalog number: 077124.09
also known as 077124-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5593-70-4
IUPAC Name
titanium(4+) tetrakis(butan-1-olate)
Molecular Formula
C16H36O4Ti
InChI Key
YHWCPXVTRSHPNY-UHFFFAOYSA-N
SMILES
[Ti+4].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-]
Appearance (Color)
Clear colorless to yellow to orange to pale brown
Form
Liquid
Chloride content
≤50 ppm (U.S. specification)
Assay from Suppliers CofA
Titanium Dioxide (TiO2): ≥23.0% (U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

Titanium(IV) n-butoxide is used in the preparation of nanosized titania powders in the anatase form and ferroelectric bismuth titanate thin films. It is also used as intermediates, paint additives, coating additives, processing aids and as process regulators. It is involved in the preparation of nanocrystalline titanium dioxide powders at room temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text