Titanium(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Titanium(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 077126.06
also known as 077126-06
Price (USD)/ Each
32.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
32.00
Add to cart
Titanium(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number077126.06
Price (USD)/ Each
32.00
-
Add to cart
Chemical Identifiers
CAS12137-20-1
IUPAC Nameoxotitanium
Molecular FormulaOTi
InChI KeyOGIDPMRJRNCKJF-UHFFFAOYSA-N
SMILESO=[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Titanium(II) oxide associated with vanadium oxide forms a transition-metal-oxide monolayer catalyst, which is used in the selective oxidation and ammoxidation of alkylaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only