Thermo Scientific Chemicals

Titanium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals

Catalog number: 077130.09
10 g, Each
Thermo Scientific Chemicals

Titanium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals

Catalog number: 077130.09
10 g, Each
Quantity
Catalog number: 077130.09
also known as 077130-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14024-64-7
IUPAC Name
bis(4-hydroxypent-3-en-2-one); oxotitanium
Molecular Formula
C10H16O5Ti
InChI Key
ADVORQMAWLEPOI-UHFFFAOYSA-N
SMILES
O=[Ti].CC(O)=CC(C)=O.CC(O)=CC(C)=O
Assay from Supplier's CofA
≥98.0% (titration)
Form
Crystals or crystalline powder
Appearance (Color)
Pale yellow

Description

Employed as a cross linking agent for cellulose fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text