Thermo Scientific Chemicals

Titanium(IV) ethoxide, TiO{2} 33% min, Thermo Scientific Chemicals

Catalog number: 077142.30
250 g, Each
Thermo Scientific Chemicals

Titanium(IV) ethoxide, TiO{2} 33% min, Thermo Scientific Chemicals

Catalog number: 077142.30
250 g, Each
Quantity
Catalog number: 077142.30
also known as 077142-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3087-36-3
IUPAC Name
titanium(4+) tetraethanolate
Molecular Formula
C8H20O4Ti
InChI Key
JMXKSZRRTHPKDL-UHFFFAOYSA-N
SMILES
[Ti+4].CC[O-].CC[O-].CC[O-].CC[O-]
Form
viscous liquid/ fused solidified melt
Assay (unspecified)
TiO₂ 33%, min
Appearance (Color)
Colorless to yellow

Description

Titanium(IV) ethoxide is used in organic synthesis and material science. It is involved in the synthesis of anti-perspirants and antifungal agents. Furthermore, it is used to prepare carbon films in association with silicon dioxide. It is also used as a precursor for the synthesis of Ti4+ ions titanium nanoparticles by various methods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text