Thermo Scientific Chemicals

Thallium(I) fluoride, 97%, Thermo Scientific Chemicals

Catalog number: 078111.22
100 g, Each
Thermo Scientific Chemicals

Thallium(I) fluoride, 97%, Thermo Scientific Chemicals

Catalog number: 078111.22
100 g, Each
Quantity
Catalog number: 078111.22
also known as 078111-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-27-7
IUPAC Name
λ¹-thallanylium fluoride
Molecular Formula
FTl
InChI Key
CULOEOTWMUCRSJ-UHFFFAOYSA-M
SMILES
[F-].[Tl+]
Assay (unspecified)
96.1% min

Description

Thallium(I) fluoride is used as chemical reagents, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text