Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Use Thermo Scientific™ MAbPac™ SCX-10 LC Columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants. The unique nonporous pellicular resin provides the resolving power to separate mAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.
Column Format | Analytical Column |
Diameter (Metric) | 4 mm |
Flow Rate | 2 mL/min |
Length (Metric) | 50 mm |
Max. Pressure | 5000 psi (345 bar) |
Particle Size | 5 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Solvents | 50% Acetonitrile |
Temperature | Ambient to 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Material | PEEK |
Type | Analytical Column |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 4 mm |
Flow Rate | 1 mL/min |
Length (Metric) | 250 mm |
Max. Pressure | 3000 psi (207 bar) |
Particle Size | 10 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Solvents | 50% Acetonitrile |
Temperature | Ambient to 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Type | Analytical Column |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 4 mm |
Flow Rate | 0.2 to 2.0 mL/min |
Length (Metric) | 150 mm |
Max. Pressure | 3000 psi (207 bar) |
Particle Size | 10 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Solvents | 50% Acetonitrile |
Temperature | Ambient to 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Type | Analytical Column |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Preparative Column, Standard |
Diameter (Metric) | 22 mm |
Length (Metric) | 250 mm |
Max. Pressure | 3000 psi (207 bar) |
Particle Size | 10 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | Ambient to 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Technique | Monoclonal Antibody/Cation Exchange |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 4 mm |
Flow Rate | 0.2 to 2.0 mL/min |
Length (Metric) | 50 mm |
Max. Pressure | 3000 psi (207 bar) |
Particle Size | 10 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Solvents | 50% Acetonitrile |
Temperature | Ambient to 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Type | Analytical Column |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 2 mm |
Flow Rate | 0.25 mL/min |
Length (Metric) | 250 mm |
Max. Pressure | 3000 psi (207 bar) |
Particle Size | 10 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Solvents | 50% Acetonitrile |
Temperature | Ambient to 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Type | Analytical Column |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 4 mm |
Length (Metric) | 150 mm |
Max. Pressure | 5000 psi (345 bar) |
Particle Size | 5 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | Ambient to 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Type | Analytical Column |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 4.6 mm |
Flow Rate | 2 mL/min |
Length (Metric) | 50 mm |
Max. Pressure | 7000 psi (480 bar) |
Particle Size | 5 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | Ambient to 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Type | Analytical Column |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 4.6 mm |
Flow Rate | 1.3 mL/min |
Functionality/Interaction Type | Sulfonic |
Length (Metric) | 150 mm |
Max. Pressure | 7000 psi (480 bar) |
Particle Size | 5 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | Ambient to 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Type | Analytical Column |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 4.6 mm |
Flow Rate | 0.8 mL/min |
Functionality/Interaction Type | Sulfonic |
Length (Metric) | 250 mm |
Max. Pressure | 7000 psi (480 bar) |
Particle Size | 5 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | Ambient to 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Type | AnalyticalColumn |
Technique | Bio-LC |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Particle Size | Diameter (Metric) | Length (Metric) | Column Format | Price (USD) | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
078656 | Each | 5 μm | 4 mm | 50 mm | Analytical Column | 1,117.80 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
074625 | Each | 10 μm | 4 mm | 250 mm | Analytical Column | 1,481.79 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
075602 | Each | 10 μm | 4 mm | 150 mm | Analytical Column | 1,315.06 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SP6947 | Each | 10 μm | 22 mm | 250 mm | Preparative Column, Standard | Request A Quote | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
075603TS also known as 075603 | Each | 10 μm | 4 mm | 50 mm | Analytical Column | 1,094.77 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
075604 | Each | 10 μm | 2 mm | 250 mm | Analytical Column | 1,481.79 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
085198 | Each | 5 μm | 4 mm | 150 mm | Analytical Column | 1,816.43 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
082674 | Each | 5 μm | 4.6 mm | 50 mm | Analytical Column | 2,273.18 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
085209 | Each | 5 μm | 4.6 mm | 150 mm | Analytical Column | 2,728.75 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
082673 | Each | 5 μm | 4.6 mm | 250 mm | Analytical Column | 3,031.69 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|