Vanadium(III) fluoride, 98%, Thermo Scientific Chemicals
Vanadium(III) fluoride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium(III) fluoride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 081115.04
also known as 081115-04
Price (USD)/ Each
63.50
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
63.50
Add to cart
Vanadium(III) fluoride, 98%, Thermo Scientific Chemicals
Catalog number081115.04
Price (USD)/ Each
63.50
-
Add to cart
Chemical Identifiers
CAS10049-12-4
IUPAC Namevanadium(3+) trifluoride
Molecular FormulaF3V
InChI KeyPASVSMBJEYQYSN-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[V+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.5% min
Employed in oxygen-sensitive applications, such as metal production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only