Thermo Scientific Chemicals

Vanadium(V) trichloride oxide, V+5 28.5% min, Thermo Scientific Chemicals

Catalog number: 081118.09
10 g, Each
Thermo Scientific Chemicals

Vanadium(V) trichloride oxide, V+5 28.5% min, Thermo Scientific Chemicals

Catalog number: 081118.09
10 g, Each
Quantity
Catalog number: 081118.09
also known as 081118-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7727-18-6
IUPAC Name
oxovanadiumpentakis(ylium) trichloride
Molecular Formula
Cl3OV
InChI Key
WMZFXRASJAIALG-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].O=[V+5]
Assay (unspecified)
96% min

Description

Vanadium(V) trichloride oxide is used as a catalyst in olefin polymerization reaction, especially for the production of ethylene-propylene rubber. It is also used to synthesis organovanadium compounds and as a mordant in dyeing. It acts as a precursor to prepare vanadium pentoxide, vanadyl dichloride and dioxovanadium monochloride. Further, it serves as a reagent and a strong oxidizing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text