Thermo Scientific Chemicals

Vanadium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals

Catalog number: 081119.09
10 g, Each
Thermo Scientific Chemicals

Vanadium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals

Catalog number: 081119.09
10 g, Each
Quantity
Catalog number: 081119.09
also known as 081119-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3153-26-2
IUPAC Name
oxovanadiumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular Formula
C10H14O5V
InChI Key
JFHJZWAQYMGNBE-SUKNRPLKSA-L
SMILES
O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Appearance (Color)
Blue to green
Melting Point
250 to 254?C
Evaporation loss
0.30% max. volatile content
Form
Crystalline powder
Insoluble material
0.30%, max (in Dichloromethane)

Description

Employed as a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment. It is used in organic chemistry as a reagent in the epoxidation of allylic alcohols in combination with tert-butyl hydroperoxide (TBHP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text