Vanadium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals
Vanadium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals

As a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number 081119.18
also known as 081119-18
Price (USD)/ Each
68.65
Online exclusive
75.80 
Save 7.15 (9%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
68.65
Online exclusive
75.80 
Save 7.15 (9%)
Add to cart
Vanadium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals
Catalog number081119.18
Price (USD)/ Each
68.65
Online exclusive
75.80 
Save 7.15 (9%)
-
Add to cart
Chemical Identifiers
CAS3153-26-2
IUPAC Nameoxovanadiumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14O5V
InChI KeyJFHJZWAQYMGNBE-SUKNRPLKSA-L
SMILESO=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue to green
Melting Point250 to 254?C
Evaporation loss0.30% max. volatile content
FormCrystalline powder
Insoluble material0.30%, max (in Dichloromethane)
Employed as a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment. It is used in organic chemistry as a reagent in the epoxidation of allylic alcohols in combination with tert-butyl hydroperoxide (TBHP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only