Thermo Scientific Chemicals

Tungstic(VI) acid, Thermo Scientific Chemicals

Catalog number: 082118.22
100 g, Each
Thermo Scientific Chemicals

Tungstic(VI) acid, Thermo Scientific Chemicals

Catalog number: 082118.22
100 g, Each
Quantity
Catalog number: 082118.22
also known as 082118-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-03-1
IUPAC Name
dioxotungstendiol
Molecular Formula
H2O4W
InChI Key
CMPGARWFYBADJI-UHFFFAOYSA-L
SMILES
O[W](O)(=O)=O
Impurity content
1% max

Description

Applied as flavoring agent and fragrance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text