Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants, use Thermo Scientific™ MAbPac™ SCX-10 RS Monoclonal Antibody Analysis Columns, 5μm. The unique nonporous pellicular resin provides the resolving power to separate mAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.
Column Format | Analytical Column |
Diameter (Metric) | 2.1 mm |
Flow Rate | 0.28 mL/min |
Length (Metric) | 250 mm |
Max. Pressure | 7000 psi (480 bar) |
Particle Size | 5 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 2.1 mm |
Flow Rate | 0.46 mL/min |
Length (Metric) | 150 mm |
Max. Pressure | 7000 psi (480 bar) |
Particle Size | 5 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | Ambient to 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Technique | Bio-LC |
Unit Size | Each |
Column Format | Analytical Column |
Diameter (Metric) | 2.1 mm |
Flow Rate | 0.5 mL/min |
Length (Metric) | 50 mm |
Max. Pressure | 7000 psi (480 bar) |
Particle Size | 5 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | Ambient to 60°C |
Stationary Phase | Sulfonate |
Product Line | MAbPac |
Packing Material | Spherical, Nonporous Polymer |
Column Type | Ion Exchange |
Phase | Ion Exchange |
Technique | Bio-LC |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Particle Size | Diameter (Metric) | Length (Metric) | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
082515 | Each | 5 μm | 2.1 mm | 250 mm | 3,031.69 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
088242 | Each | 5 μm | 2.1 mm | 150 mm | 2,728.75 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
082675 | Each | 5 μm | 2.1 mm | 50 mm | 2,273.18 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|