T-1270 Fixed Angle Titanium Rotor
T-1270 Fixed Angle Titanium Rotor
Thermo Scientific™

T-1270 Fixed Angle Titanium Rotor

Effectively perform a wide range of applications with the T-1270 Rotor, which offers 12-place capacity.
Have Questions?
Catalog NumberCapacity
0825912 x 12.5 mL
Catalog number 08259
Price (EUR)
-
Request A Quote
Capacity:
12 x 12.5 mL
Ask our AI about this Product

Effectively perform a wide range of applications with the Thermo Scientific™ T-1270 Fixed Angle Rotor, which offers 12-place capacity.

Recommended for:

DNA isolation, serum flotation.

Specifications
Angle24°
Avg. RCF335,513 x g
DescriptionT-1270 Fixed Angle Titanium Rotor
For Use With (Equipment)Thermo Scientific™ Sorvall™ WX ultraCentrifuges
K Factor36.3
Max. Tube Diameter16 mm
Max. Tube Length80 mm
Min. RCF222,216 x g
Net Weight13 lb. (5.9 kg)
Radius (Metric)82 mm
Radius (Metric) Min.40.6 mm
Capacity12 x 12.5 mL
MaterialTitanium
Max. RCF448,811 x g
Max. Speed70,000 rpm
TypeFixed Angle Rotor
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection