1x1 image pixel for data collection
Acclaim™ SEC-300 Size Exclusion Chromatography HPLC Columns
Product Image
Thermo Scientific™

Acclaim™ SEC-300 Size Exclusion Chromatography HPLC Columns

Green features

Separate water-soluble polymers and oligomers with the Acclaim family of resin-based, high-performance, size-exclusion chromatography columns.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
08274033 mm4.6 mm
079723300 mm4.6 mm
079725300 mm7.8 mm
079726150 mm7.8 mm
Catalog number 082740
Price (USD)
912.48
Each
Add to cart
Length (Metric):
33 mm
Diameter (Metric):
4.6 mm
Price (USD)
912.48
Each
Add to cart

Separate water-soluble polymers and oligomers with Thermo Scientific™ Acclaim™ SEC-300 Size Exclusion Chromatography HPLC Columns. These resin-based, high-performance, size-exclusion chromatography columns cover molecular weight range: 5μm, 300Å. The combination of a proprietary mono-dispersed multi-pore hydrophilic resin and small particle size provides high-resolution and good linearity of the calibration curve with no inflection points for accurate, reliable analysis of a variety of water-soluble polymers.

The Acclaim SEC-300 column is calibrated from 100 to 50,000 Daltons and offers good resolution for lower molecular weight polymers or oligomers.

High-Performance Analysis of Water-Soluble Polymers
  • High-resolution analysis at reduced solvent consumption in available 4.6 x 300mm dimension
  • Stable surface bonding leads to low column bleed and enhances compatibility with UV, RI, MS, ELSD and Thermo Scientific Dionex™ Corona™ Charged Aerosol Detectors
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatGuard Column
Column TypeSize Exclusion
EndcappedYes
Green FeaturesResponsibly packaged
Packing MaterialResin
Particle ShapeSpherical
QuantityEach
Stationary PhaseProprietary
Surface Area1000 m²/g
Temperature60°C
USP TypeN/A
Diameter (Metric)4.6 mm
Length (Metric)33 mm
Particle Size5 μm
PhaseSize Exclusion
Pore Size300 Å
Product LineAcclaim
TechniqueBio-LC
pH2 to 12
Unit SizeEach

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link