pH Gradient Buffer Concentrates
pH Gradient Buffer Concentrates
Actual product may vary
Thermo Scientific™

pH Gradient Buffer Concentrates

Robust reproducible gradients that are applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionpHQuantity
083273CX-1 pH Gradient Buffer A, 10X Concentrate5.6125 mL
083275CX-1 pH Gradient Buffer B, 10X Concentrate10.2125 mL
085346CX-1 pH Gradient Buffer A, 10X Concentrate5.6250 mL
085348CX-1 pH Gradient Buffer B, 10X Concentrate10.2250 mL
302779CX-1 pH Gradient Buffer A, 10X Concentrate5.6500 mL
302780CX-1 pH Gradient Buffer B, 10X Concentrate10.2500 mL
303274CX-1 pH Gradient Buffer A, 10X Concentrate5.61 L
303275CX-1 pH Gradient Buffer B, 10X Concentrate10.21 L
Catalog number 083273
Price (USD)
217.73
Each
Add to cart
Description:
CX-1 pH Gradient Buffer A, 10X Concentrate
pH:
5.6
Quantity:
125 mL
Price (USD)
217.73
Each
Add to cart

Obtain fast and highly robust, reproducible HPLC gradients using Thermo Scientific™ CX-1 pH Gradient Buffers. Simple to optimize and easily automated, these buffers are applicable for HPLC separation of the majority of monoclonal antibody (mAb) biotherapeutics drugs. CX-1 pH Gradient Buffers are ready to use with existing HPLC columns without the need to formulate mobile phases. The CX-1 buffers save time in method development, facilitate method transfer to QA/QC for a wide range of mAb charge variants through a generic HPLC-based approach to mAb characterization, and do not require time-consuming mobile phase adjustments.

pH buffer solutions can be used to generate highly reproducible, linear pH gradients in cation exchange chromatography.

  • Robust, reproducible gradients
  • Predict biomolecule pH
  • Simple method optimization
  • Easily automated
  • Applicable to the majority of MAbs

Unlike traditional cation exchange chromatography using salt gradients, it is possible to predict the pH and the expected retention of the charge variants and use a narrow pH range to get a higher resolution separation.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
DescriptionCX-1 pH Gradient Buffer A, 10X Concentrate
Product LineThermo Scientific
Quantity125 mL
Volume (Metric)125 mL
For Use With (Equipment)LC Columns and Systems
TypeCX-1 pH Gradient Buffer A
pH5.6
Unit SizeEach

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link