Zirconium dichloride oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Zirconium dichloride oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium dichloride oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 086108.30
Price (USD)/ Each
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS15461-27-5
IUPAC Namedichloro(oxo)zirconium
Molecular FormulaCl2OZr
InChI KeyIPCAPQRVQMIMAN-UHFFFAOYSA-L
SMILESCl[Zr](Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds an application as rubber additive and fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only