Thermo Scientific Chemicals

Zirconium(IV) fluoride, 98%, Thermo Scientific Chemicals

Catalog number: 086112.30
250 g, Each
Thermo Scientific Chemicals

Zirconium(IV) fluoride, 98%, Thermo Scientific Chemicals

Catalog number: 086112.30
250 g, Each
Quantity
Catalog number: 086112.30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7783-64-4
IUPAC Name
zirconium(4+) tetrafluoride
Molecular Formula
F4Zr
InChI Key
OMQSJNWFFJOIMO-UHFFFAOYSA-J
SMILES
[F-].[F-].[F-].[F-].[Zr+4]
Assay (unspecified)
97.6% min

Description

Zirconium(IV) fluoride is used as a zirconium source in oxygen-sensitive applications such as metal production. Together with uranium salt, zirconium fluoride can be a component of fuel-coolant in molten salt reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text