Thermo Scientific Chemicals

Zirconium(IV) isopropoxide isopropanol complex, Thermo Scientific Chemicals

Catalog number: 086118.18
50 g, Each
Thermo Scientific Chemicals

Zirconium(IV) isopropoxide isopropanol complex, Thermo Scientific Chemicals

Catalog number: 086118.18
50 g, Each
Quantity
Catalog number: 086118.18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14717-56-7
IUPAC Name
zirconium(4+) propan-2-ol tetrakis(propan-2-olate)
Molecular Formula
C15H36O5Zr
InChI Key
NIJDLRJGRCHJDB-UHFFFAOYSA-N
SMILES
[Zr+4].CC(C)O.CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Elemental analysis by ICP
Cu <10 ppm
Elemental analysis by ICP
Hf <200 ppm
Elemental analysis by ICP
Mg <10 ppm
Elemental analysis by ICP
Ti <50 ppm
Elemental analysis by ICP
V <10 ppm

Description

Zirconium(IV) isopropoxide isopropanol is a suitable precursor for the synthesis of olefin-functionalized zirconium containing monomers. It is used in the manufacture of hybrid zirconium containing polymers. It is also used as a source of zirconia, a well known material having high mechanical strength, chemical durability and alkali resistance. It is also used for the chemical protection of metal substrates from acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text