Thermo Scientific Chemicals

Zirconium(IV) bromide, 99%, Thermo Scientific Chemicals

Catalog number: 086139.18
50 g, Each
Thermo Scientific Chemicals

Zirconium(IV) bromide, 99%, Thermo Scientific Chemicals

Catalog number: 086139.18
50 g, Each
Quantity
Catalog number: 086139.18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13777-25-8
Assay (unspecified)
98.6% min

Description

Zirconium(IV) bromide is used as a starting material in the synthesis of zirconium hydride complexes as well as unique cluster compounds. It is also used as a biochemical for proteomics research and as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text