Thermo Scientific Chemicals

Ferrocene, 99%, Thermo Scientific Chemicals

Catalog number: 087202.30
250 g, Each
Thermo Scientific Chemicals

Ferrocene, 99%, Thermo Scientific Chemicals

Catalog number: 087202.30
250 g, Each
Quantity
Catalog number: 087202.30
also known as 087202-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
102-54-5
IUPAC Name
Ferrocene
Molecular Formula
C10H10Fe
InChI Key
DFRHTHSZMBROSH-UHFFFAOYSA-N
SMILES
[Fe].c1cccc1.c1cccc1
Assay (GC)
≥98.5%
Appearance (Color)
Yellow to orange
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Elemental analysis by ICP
Fe (Iron): 28.0-33.0%

Description

Ferrocene is used as a catalyst for carbon nanotubes and for rocket propellants. It is also used in the fuel for petrol engines, gasoline antiknock, luminous sensitive materials and exhibit anticancer activity. It acts as a photosensitive catalyst and promote the photochemical degadation of plastic films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text