Ferrocene, 99%, Thermo Scientific Chemicals
Ferrocene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ferrocene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Catalog number 087202.A1
also known as 087202-A1
Price (USD)/ Each
239.65
Online exclusive
266.00 
Save 26.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
239.65
Online exclusive
266.00 
Save 26.35 (10%)
Add to cart
Ferrocene, 99%, Thermo Scientific Chemicals
Catalog number087202.A1
Price (USD)/ Each
239.65
Online exclusive
266.00 
Save 26.35 (10%)
-
Add to cart
Chemical Identifiers
CAS102-54-5
IUPAC NameFerrocene
Molecular FormulaC10H10Fe
InChI KeyDFRHTHSZMBROSH-UHFFFAOYSA-N
SMILES[Fe].c1cccc1.c1cccc1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange or orange-brown
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Elemental analysis by ICPFe (Iron): 28.0-33.0%
View more
Ferrocene is used as a catalyst for carbon nanotubes and for rocket propellants. It is also used in the fuel for petrol engines, gasoline antiknock, luminous sensitive materials and exhibit anticancer activity. It acts as a photosensitive catalyst and promote the photochemical degadation of plastic films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only