Copper(II) cyclohexanebutyrate, AAS, Cu 15.8%, Thermo Scientific Chemicals
Catalog number087262.06
Price (USD)/ Each
175.00
-
Add to cart
Specifications
Chemical Name or MaterialCopper (II) cyclohexanebutyrate
CAS2218-80-6
Melting Point126°C (decomposition)
Recommended StorageAmbient temperatures
EINECS Number218-723-7
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.