Ethylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals
Ethylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals

Grignard reagent
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mol
1 mol
Catalog number 087291.JT
also known as 087291-JT
Price (USD)/ Each
59.65
Online exclusive
65.80 
Save 6.15 (9%)
-
Add to cart
Quantity:
0.5 mol
Request bulk or custom format
Price (USD)/ Each
59.65
Online exclusive
65.80 
Save 6.15 (9%)
Add to cart
Ethylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals
Catalog number087291.JT
Price (USD)/ Each
59.65
Online exclusive
65.80 
Save 6.15 (9%)
-
Add to cart
Chemical Identifiers
CAS925-90-6
IUPAC Namebromo(ethyl)magnesium
Molecular FormulaC2H5BrMg
InChI KeyTWTWFMUQSOFTRN-UHFFFAOYSA-M
SMILESCC[Mg]Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to faintly turbid
FormLiquid
Concentration2.85-3.30M
Ethylmagnesium bromide is used as grignard reagent. It is widely used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only