Thermo Scientific Chemicals

Benzylmagnesium chloride, 1-2M in THF, Thermo Scientific Chemicals

Catalog number: 087299.AD
50 mL, Each
Thermo Scientific Chemicals

Benzylmagnesium chloride, 1-2M in THF, Thermo Scientific Chemicals

Catalog number: 087299.AD
50 mL, Each
Quantity
Catalog number: 087299.AD
also known as 087299-AD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6921-34-2
IUPAC Name
benzyl(chloro)magnesium
Molecular Formula
C7H7ClMg
InChI Key
NRAFPLGJPPJUNB-UHFFFAOYSA-M
SMILES
Cl[Mg]CC1=CC=CC=C1
Appearance (Color)
Pale brown to dark brown / purple
Form
Hazy liquid
Assay (Aqueous acid-base Titration)
1-2M

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text