Nickel(II) oxide, black, Ni 76%, (contains Ni{2}O{3}), Thermo Scientific Chemicals
Nickel(II) oxide, black, Ni 76%, (contains Ni{2}O{3}), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) oxide, black, Ni 76%, (contains Ni{2}O{3}), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 087302.A1
also known as 087302-A1
Price (USD)/ Each
230.65
Online exclusive
256.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
230.65
Online exclusive
256.00 
Save 25.35 (10%)
Add to cart
Nickel(II) oxide, black, Ni 76%, (contains Ni{2}O{3}), Thermo Scientific Chemicals
Catalog number087302.A1
Price (USD)/ Each
230.65
Online exclusive
256.00 
Save 25.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1313-99-1
IUPAC Nameoxonickel
Molecular FormulaNiO
InChI KeyGNRSAWUEBMWBQH-UHFFFAOYSA-N
SMILESO=[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)76% Ni min
Nickel(II) oxide is used in fuel cell electrodes, storage batteries, electronic circuit boards, electrical ceramics, and as a pigment for ceramics and glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only