Cobalt(II) benzoate, Thermo Scientific Chemicals
Cobalt(II) benzoate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) benzoate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 087380.09
also known as 087380-09
Price (USD)/ Each
38.40
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
38.40
Add to cart
Cobalt(II) benzoate, Thermo Scientific Chemicals
Catalog number087380.09
Price (USD)/ Each
38.40
-
Add to cart
Chemical Identifiers
CAS932-69-4
IUPAC Nameλ²-cobalt(2+) dibenzoate
Molecular FormulaC14H10CoO4
InChI KeyGAYAMOAYBXKUII-UHFFFAOYSA-L
SMILES[Co++].[O-]C(=O)C1=CC=CC=C1.[O-]C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
FormCrystals or crystalline powder
Assay (unspecified)≥99.0%
Melting Point114-116°C
Cobalt(II) benzoate is used as ingredients of many cosmetic products, industrial fields such as automobile, fuel, battery, paint, metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only