Thermo Scientific Chemicals

Sodium hydrogen sulfide, anhydrous, Thermo Scientific Chemicals

Catalog number: 087389.04
2 g, Each
Thermo Scientific Chemicals

Sodium hydrogen sulfide, anhydrous, Thermo Scientific Chemicals

Catalog number: 087389.04
2 g, Each
Quantity
Catalog number: 087389.04
also known as 087389-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16721-80-5
IUPAC Name
sodium sulfanide
Molecular Formula
HNaS
InChI Key
HYHCSLBZRBJJCH-UHFFFAOYSA-M
SMILES
[Na+].[SH-]
Assay (unspecified)
38.5% Na min

Description

In paper pulping, in dyestuffs processing, in rayon and cellophane desulfurizing, in unhairing hides and as a bleaching reagent Sodium hydrogen sulfide is used in cloth, paper, pulp and dyestuff industries. It acts as a floating agent in copper mining as well as to remove hairs from hides in leather industries. It is also used as an intermediate in synthetic chemistry. It is utilized as a raw material in the production of ammonium sulphate and ethanethiol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Moisture Sensitive and Hygroscopic
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text