Thermo Scientific Chemicals

Sodium bromide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 087604.14
25 g, Each
Thermo Scientific Chemicals

Sodium bromide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 087604.14
25 g, Each
Quantity
Catalog number: 087604.14
also known as 087604-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7647-15-6
IUPAC Name
sodium bromide
Molecular Formula
BrNa
InChI Key
JHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES
[Na+].[Br-]
Total Metal Impurities
0.01% max.

Description

Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text