Sodium chloride, 99.99% (metals basis), Thermo Scientific Chemicals
Sodium chloride, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium chloride, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 087605.18
also known as 087605-18
Price (USD)/ Each
76.65
Online exclusive
84.60 
Save 7.95 (9%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
76.65
Online exclusive
84.60 
Save 7.95 (9%)
Add to cart
Sodium chloride, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number087605.18
Price (USD)/ Each
76.65
Online exclusive
84.60 
Save 7.95 (9%)
-
Add to cart
Chemical Identifiers
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (special)?2.5 % of Theoretical value (Na)
Total Metal Impurities≤0.01% (100 ppm)
FormCrystals
Sodium chloride is widely used as a food additive, and a preservative. It is employed in the Chloralkali process for producing chlorine and sodium hydroxide, in the Solvay process for producing sodium carbonate and calcium chloride, in the Mannheim process and in the Hargreaves process for producing sodium sulfate and hydrochloric acid. It is used in the manufacture of sodium chlorate, water softening and de-icing of roadways. Further it is used in several industries like pharmaceutical, textile, dyeing, rubber food and agriculture. It is used in the processing of steel, copper, vanadium, and other metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only