Sodium sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
Sodium sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 087611.22
also known as 087611-22
Price (USD)/ Each
561.65
Online exclusive
624.00 
Save 62.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
561.65
Online exclusive
624.00 
Save 62.35 (10%)
Add to cart
Sodium sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number087611.22
Price (USD)/ Each
561.65
Online exclusive
624.00 
Save 62.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7757-82-6
IUPAC Namedisodium sulfate
Molecular FormulaNa2O4S
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only