Bromine liquid, 99.99+% (metals basis), Thermo Scientific Chemicals
Bromine liquid, 99.99+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromine liquid, 99.99+% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
5 mL
250 mL
Catalog number 087614.AD
also known as 087614-AD
Price (USD)/ Each
246.00
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
246.00
Add to cart
Bromine liquid, 99.99+% (metals basis), Thermo Scientific Chemicals
Catalog number087614.AD
Price (USD)/ Each
246.00
-
Add to cart
Chemical Identifiers
CAS7726-95-6
IUPAC Namebromide
Molecular FormulaBr
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-M
SMILES[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Very Dark Red-Brown
FormLiquid
Assay from Supplier's CofAFree Chlorine ≤ 0.05 %
Assay from Supplier's CofAIodine ≤ 0.001 %
Assay from Supplier's CofANickel (Ni) ≤ 5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only