Cesium bromide, 99.999% (metals basis), Thermo Scientific Chemicals
Cesium bromide, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cesium bromide, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 087639.09
also known as 087639-09
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
Add to cart
Cesium bromide, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number087639.09
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7787-69-1
IUPAC Namecaesium(1+) bromide
Molecular FormulaBrCs
InChI KeyLYQFWZFBNBDLEO-UHFFFAOYSA-M
SMILES[Br-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline
Particle Size-4 Mesh
Trace Metals (by ICP)≤0.001% (10 ppm)
Cesium bromide is a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only