Thermo Scientific Chemicals

Cesium bromide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 087639.18
50 g, Each
Thermo Scientific Chemicals

Cesium bromide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 087639.18
50 g, Each
Quantity
Catalog number: 087639.18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7787-69-1
IUPAC Name
caesium(1+) bromide
Molecular Formula
BrCs
InChI Key
LYQFWZFBNBDLEO-UHFFFAOYSA-M
SMILES
[Br-].[Cs+]
Appearance (Color)
White
Form
Crystalline
Particle Size
-4 Mesh
Trace Metals (by ICP)
≤0.001% (10 ppm)

Description

Cesium bromide is a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text