Thermo Scientific Chemicals

Antimony(III) acetate, 97%, Thermo Scientific Chemicals

Catalog number: 087657.09
10 g, Each
Thermo Scientific Chemicals

Antimony(III) acetate, 97%, Thermo Scientific Chemicals

Catalog number: 087657.09
10 g, Each
Quantity
Catalog number: 087657.09
also known as 087657-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6923-52-0
Appearance (Color)
White to pale cream
Chloride content
< 20 ppm
Elemental Analysis
% Sb 40 - 42
Trace Metals
< 20 ppm Fe
Form
crystalline powder

Description

Antimony(III) acetate is a catalyst used in the production of synthetic fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text