Thermo Scientific Chemicals

Sodium borohydride, stable aq. soln., 4.4M in 14M NaOH, Thermo Scientific Chemicals

Catalog number: 087659.AE
100 mL, Each
Thermo Scientific Chemicals

Sodium borohydride, stable aq. soln., 4.4M in 14M NaOH, Thermo Scientific Chemicals

Catalog number: 087659.AE
100 mL, Each
Quantity
Catalog number: 087659.AE
also known as 087659-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16940-66-2
IUPAC Name
sodium boranuide
Molecular Formula
BH4Na
InChI Key
YOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES
[BH4-].[Na+]
Assay (unspecified)
NaBH₄ 11.85û12.15%
Assay (unspecified)
NaOH 38.00-42.00%

Description

Sodium borohydride is used as a reagent in the reduction of amino acids and their derivatives. Also used as a catalyst in ammonia borane dehydrogenation. It acts as a reducing agent in the synthesis of gold nanoparticles. It is also used for wood pulp and as a blowing agent for plastics. It reduces aldehyde and keto groups to give the corresponding alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text