Thermo Scientific Chemicals

Iodine(V) oxide, 98%, Thermo Scientific Chemicals

Catalog number: 087682.22
100 g, Each
Thermo Scientific Chemicals

Iodine(V) oxide, 98%, Thermo Scientific Chemicals

Catalog number: 087682.22
100 g, Each
Quantity
Catalog number: 087682.22
also known as 087682-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12029-98-0
IUPAC Name
iodyl iodate
Molecular Formula
I2O5
InChI Key
BIZCJSDBWZTASZ-UHFFFAOYSA-N
SMILES
O=[I](=O)O[I](=O)=O
Appearance (Color)
White to cream or pale yellow
Form
Crystals or powder or crystalline powder
Assay (Iodometric Titration)
≥97.5 to ≤102.5%

Description

Iodine (V) oxide is used as an oxidizing agent to remove carbon monoxide from gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text