Thermo Scientific Chemicals

Cobalt(II) oxalate dihydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 087758.22
100 g, Each
Thermo Scientific Chemicals

Cobalt(II) oxalate dihydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 087758.22
100 g, Each
Quantity
Catalog number: 087758.22
also known as 087758-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5965-38-8
IUPAC Name
(5Z)-5-[(3-chloro-4-hydroxy-5-methoxyphenyl)methylidene]-1-(prop-2-en-1-yl)-2-sulfanylidene-1,3-diazinane-4,6-dione
Molecular Formula
C15H13ClN2O4S
InChI Key
YOKXELRIGMPSIG-UITAMQMPSA-N
SMILES
COC1=CC(C=C2C(=O)NC(=S)N(CC=C)C2=O)=CC(Cl)=C1O
Appearance (Color)
White to pink
Assay (unspecified)
Co: ≥31.2%
Identification (FTIR)
Conforms
Form
Powder

Description

Cobalt(II) oxalate dihydrate is used as a stabilizer for hydrogen cyanide and as a temperature indicator. It is used in the preparation of cobalt catalysts, and cobalt metal powder for powder-metallurgical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text